Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pepstatin A, BODIPY™ FL Conjugate

Catalog No. P12271
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12271 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12271 Supplier Invitrogen™ Supplier No. P12271
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For the study of cathepsin D trafficking in cells by fluorescence microscopy

BODIPY™ FL pepstatin A, a green-fluorescent probe, which inhibits cathepsin D in vitro (IC50 approx. 50nM), targets cathepsin D within lysosomes of live cells. Initial studies with this new probe indicate that it is transported to the lysosomes of live cells via an endocytic pathway and that it can be competitively displaced by unlabeled pepstatin A.

Apoptosis, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 μg
Shipping Condition Room Temperature
Product Line BODIPY
Physical Form Solid
Product Type Inhibitor
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.