Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qtracker™ 565 Vascular Labels

Catalog No. Q21031MP
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q21031MP 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q21031MP Supplier Invitrogen™ Supplier No. Q21031MP
  • $832.00
    $682.24 / Each of 1

    Save $149.76

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

200μL Qtracker non-targeted quantum dots (2μM solution in 50mm borate buffer, pH 8.3).

Non-targeted quantum dots are designed to be injected into tail vein of mice for study of vascular structure using small animal in vivo imaging (SAIVI) techniques

  • These nanocrystals exhibit intense fluorescence with red-shifted emission for increased tissue penetration, and have PEG surface coating specially developed to minimize nonspecific interactions and reduce immune response by tissue
  • Because the PEG surface coating does not contain reactive functional groups, dots are retained in circulation longer and can be imaged for up to 3 hours with single injection or for longer periods of time with additional injections
  • Qtracker 565 label has Ex/Em (405-525/565)nm

Cell Analysis, Cellular Imaging, In Vivo Imaging

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 μM
Content And Storage Contains 200 μl Qtracker™ non-targeted quantum dots (2 μM solution in 50 mM borate buffer, pH 8.3). Store at 2–6°C. Do not freeze. Stable for at least 6 months.
Dye Type Qdot Nanocrystals
Quantity 200 μL
Product Line QTRACKER, Qdot
Reagent Type Vascular Imaging Reagents
Shipping Condition Room Temperature
Technique In Vivo Imaging
Product Type Vascular Label
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.