Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Secure-Seal™ Hybridization Chamber Gasket, 8 chambers, 9 mm diameter, 0.8 mm deep

Catalog No. S24732
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24732 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24732 Supplier Invitrogen™ Supplier No. S24732
  • $262.00
    $214.84 / Each of 1

    Save $47.16

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Coverslip-seal combination that attaches to microscope slides to form microwells optimized for carrying out in situ hybridization procedures

  • Special adhesive that bonds to glass slides in seconds, creating water-tight seal that is temperature resistant but can also be removed cleanly and easily after hybridization
  • Solutions are easily added or removed through dual-access ports without disrupting specimen
  • Hydrophobic coverslips are nuclease-free and will not trap or bind probes, allowing uniform distribution of reagent over specimen

Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, Gene Expression Analysis and Genotyping, In Situ Hybridization (ISH), Microarray Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Hybridization Chamber Gasket
Product Line Secure-Seal
Depth (Metric) 0.8 mm
Diameter (Metric) 9 mm
Content And Storage Store at room temperature.
Quantity 1 set
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.