Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sulforhodamine B

Catalog No. S1307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S1307 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1307 Supplier Invitrogen™ Supplier No. S1307
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble, nonfixable red fluorescent (excitation/emission maxima approximately 565/586nm) polar tracer

  • Can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion

Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 5 g
Detection Method Fluorescence
Shipping Condition Room Temperature
Product Type Sulforhodamine
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.