Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTO™ Blue Fluorescent Nucleic Acid Stain Sampler Kit (SYTO™ dyes 40, 41, 42, 45)

Catalog No. S11350
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11350 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11350 Supplier Invitrogen™ Supplier No. S11350
  • $546.00
    $447.72 / Each of 1

    Save $98.28

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

The kit contains 50μL of each SYTO dye.

Includes cell-permeant, blue-fluorescent SYTO dyes

The SYTO Blue Fluorescent Nucleic Acid Stain Sampler Kit contains a collection of cell-permeant, blue-fluorescent SYTO nucleic acid stains (SYTO dyes 40, 41, 42, 45). Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL of each SYTO dye.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage The kit contains 50 μL of each SYTO dye.
  • Store in freezer -5°C to -30°C and protect from light.
  • Excitation Wavelength Range various
    Dye Type Cell-Permeant
    For Use With (Equipment) Fluorescence Microscope
    Quantity 1 kit
    Detection Method Fluorescence
    Product Line SYTO
    Shipping Condition Room Temperature
    Label Type Fluorescent Dye
    Product Type Nucleic Acid Stain
    Sub Cellular Localization Nucleic Acids
    Show More Show Less
    How do SYTO dyes bind to DNA?

    The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

    1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
    2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
    3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
    5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

    SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.