Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tali™ Viability Kit - Dead Cell Red

Catalog No. A10786
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10786 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10786 Supplier Invitrogen™ Supplier No. A10786
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Tali - Dead Cell Red Reagent

Validated for use with the Tali Image-Based Cytometer

The TaliViability Kit - Dead Cell Red is a ready-to-use solution of propidium iodide (PI). PI is a live cell-impermeant fluorogenic DNA-binding dye that has been extensively used to identify necrotic cells. After staining a cell population using Dead Cell Red, dead cells display red fluorescence upon excitation. TaliViability Kit - Dead Cell Red can be used to identify dead cells in a population of unstained cells, or in a population of GFP-expressing cells. Compatible with eukaryotic cells.

  • Impermeant to live cells
  • Easily enters and binds to nucleic acids in necrotic cells

Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Imaging Cytometry

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Viability Kit
Cell Type Eukaryotic Cell
Detection Method Fluorescence
For Use With (Equipment) Tali™ Image-Based Cytometer
Content And Storage Store at room temperature.
Compatible Cells Eukaryotic Cells
Label or Dye Propidium Iodide
Shipping Condition Room Temperature
Format Vial
Product Line Tali
Quantity 1 Kit
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.