Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Yeast Mitochondrial Stain Sampler Kit

Catalog No. Y7530
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y7530 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Y7530 Supplier Invitrogen™ Supplier No. Y7530
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeant probes found to label mitochondria in live yeast

The Yeast Mitochondrial Stain Sampler Kit contains sample quantities of five different probes including: well characterized mitochondrial stains rhodamine 123, and DiOC6(3), rhodamine B hexyl ester, MitoTracker™ Green FM and SYTO™ 18 yeast mitochondrial stain, a proprietary green fluorescent nucleic acid stain. Each of these probes is sequestered in functioning mitochondria.

Analysis of Yeast, Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking, Microbiological Analysis, Mitochondria, Yeast Subcellular Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 1 kit
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Yeast Mitochondrial Stain Sampler Kit
Sub Cellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.