Learn More
Invitrogen™ Yeast Vacuole Marker Sampler Kit
Description
The Yeast Vacuole Marker Sampler Kit contains sample quantities of a series of novel and well established probes including: three blue-fluorescent coumarin-based stains that selectively stain the lumen of the yeast vacuole; a green-fluorescent yeast vacuole membrane marker MDY-64 for staining the vacuole membrane; and commonly used vaculate marker 5-(and-6)-carboxy-2',7-dicholorofluorescein diacetate (carboxy-DCFDA) as a standard.
Analysis of Yeast, Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking, Microbiological Analysis, Vacuoles, Yeast Subcellular Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Blue, Green |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 1 kit |
| Detection Method | Fluorescence |
| Shipping Condition | Room Temperature |
| Label Type | Fluorescent Dye |
| Product Type | Yeast Vacuole Marker Sampler Kit |
| Sub Cellular Localization | Vacuoles |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.