Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieve, 8-12 Mesh Beads, MilliporeSigma™

Catalog No. MMX1583L1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX1583L1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX1583L1 Supplier MilliporeSigma Supplier No. MX1583L1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Tan to White
Chemical Name or Material Molecular Sieve, Type 4A, 8-12 Mesh Beads
Quantity 500 g
Physical Form Solid
Molecular Formula SiO2, Na2O, Al2O3, MgO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.