Learn More
Molecular Sieve Activated Pellets, Type 3A, 1/16 in., Spectrum™ Chemical
These inert pellets allow for the separation of molecular compounds.
Supplier: Spectrum Chemical Manufacturing Corporation M12945KG
Description
- Made of inert, microporous material containing extremely small holes of uniform size to block larger molecules while allowing certain smaller molecules to pass
- Can be used in chemistry laboratory for drying reaction starting materials and separating compounds
- Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use
Specifications
| Molecular Sieve | |
| Amber Glass Bottle | |
| Pellet |
| 7631-86-9 , 1313-59-3 , 1344-28-1 , 12136-45-7 , 1309-48-4 , 14808-60-7 | |
| 5 kg |