Alfa Aesar™ Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads

Manufacturer: Alfa Aesar™ 3225630

Quantity 250g

 View more versions of this product

Catalog No. AA3225630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70955-01-0
    MDL Number MFCD00147627
    UN Number UN3077
    Chemical Name or Material Molecular sieves 4A with indicator, −8+12 (≈ 2mm) beads

    Specifications

    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 70955-01-0
    UN Number UN3077
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H317-H208
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    May cause an allergic skin reaction.
    Odor Odorless
    MDL Number MFCD00147627
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 250g
    Physical Form Beads
    Mesh Size −8+12 Mesh
    Sensitivity Hygroscopic
    Color Blue
    Chemical Name or Material Molecular sieves 4A with indicator, −8+12 (≈ 2mm) beads

    GHS H Statement
    H334-H350-H360-H317-H208
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit