Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieves, 4A, Pellets, Dia.: 3.2mm, Solstice

Catalog No. 60046246 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-246 1 kg Poly Bottle
60-046-247 5 kg Drum with inliner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-246 Supplier Solstice Supplier No. 3342941KG
Add to Cart
Add to Cart

Pellets, 3.2 mm diameter

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Brown
Quantity 1 kg
Particle Size Dia.: 3.2mm
Packaging Poly Bottle
Density 2g/cm3
Physical Form Pellets
Melting Point 400°C
Linear Formula Na12[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular sieves, 4Å
Hazard Statement P261-P305 + P351 + P338
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.