Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(III) chloride, 99.5% (metals basis)

Catalog No. p-7049335
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1403404 2 g
AA1403409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1403404 Supplier Thermo Scientific Chemicals Supplier No. 01403404
Only null left
Add to Cart

CAS: 13478-18-7 | Cl3Mo | 202.3 g/mol

Molybdenum(III) chloride reacts with tetrahydrothiophene to form complexes, which catalyze the polymerization and cyclotrimerization of alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-18-7
Molecular Formula Cl3Mo
Molecular Weight (g/mol) 202.3
MDL Number MFCD00014214
InChI Key ZSSVQAGPXAAOPV-UHFFFAOYSA-K
Synonym molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride
PubChem CID 83515
ChEBI CHEBI:30626
IUPAC Name trichloromolybdenum
SMILES Cl[Mo](Cl)Cl

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 2 g
UN Number UN3260
Solubility Information Soluble in nitric acid and sulfuric acid. Insoluble in water and dilute hydrochloric acid. Slightly soluble in ethanol and diethyl ether.
Formula Weight 202.32
Percent Purity 99.5%
Odor Acidic
Sensitivity Air and moisture sensitive
Assay (metals basis)
Chemical Name or Material Molybdenum(III) chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-775-9
RTECSNumber QA4681400
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.