Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm, Thermo Scientific Chemicals

Catalog No. p-7049123
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1297214 25 g
AA1297222 100 g
AA1297288 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1297214 Supplier Thermo Scientific Chemicals Supplier No. 01297214
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Quantity 25 g
Mesh Size −22 Mesh
Odor Odorless
Form Powder
UN Number UN3089
Percent Purity 99.9975%
Grade Puratronic
Assay (metals basis excluding W), W <1000ppm
Chemical Name or Material Molybdenum powder
Hazard Category H228-H360-H371
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-107-2
RTECSNumber QA4680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.