Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(V) chloride, 99.6% (metals basis)

Catalog No. AA1183218
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg

CAS: 10241-05-1 | Cl5Mo | 273.20 g/mol

Molybdenum pentachloride is employed as a brazing and soldering flux for monel, tungsten alloys and copper. It has been proven to be an efficient catalyst for the chlorination of aromatic rings and oxidative coupling reactions. It is a strong oxidizing agent. In the Friedel-Craft reaction, molybdenum pentachloride is used as a good lewis acid. Molybdenum pentachloride is used to prepare molybdenum by vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10241-05-1
Molecular Formula Cl5Mo
Molecular Weight (g/mol) 273.20
MDL Number MFCD00003467
InChI Key GICWIDZXWJGTCI-UHFFFAOYSA-I
Synonym molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
PubChem CID 61497
ChEBI CHEBI:30635
IUPAC Name pentachloromolybdenum
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]

Specifications

Melting Point 194°C
Color White to Gray
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.6% (metals basis)
Solubility Information Soluble in dry ether,dry alcohol,organic solvents and chlorinated solvents.
Formula Weight 273.24
Odor Pungent
Sensitivity Moisture sensitive
Appearance Black crystals
Vapor Pressure 2.3 hPa (2mm Hg) at 25°C
Chemical Name or Material Molybdenum(V) chloride
Show More Show Less
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 233-575-3
RTECSNumber QA4690000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.