Learn More
Sigma Aldrich Fine Chemicals Biosciences MolymodR molecular model s
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5444691SET
Encompass
Top quality Molymod molecular model sets are for use by students teachers and researchers. May be upgraded or customized with additional model parts for more info go to sigma-aldrich.com and enter Molymod in our search engine. Scale is 2.8 cm 100 pm (open style) and 1.45 cm 100 pm (compact semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles) bonding links and a plastic storage box.Set pictured is representative of the Student set.See our Selection Guide for detailed descriptions of set contents.
Catalog No. 50-290-1289