Learn More
Sigma Aldrich Fine Chemicals Biosciences MolyorbitalTM set Organic
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5445581SET
Encompass
The Molymod molecular orbital system uses standard shiny colored atom parts to represent the central atom cores. Average scale is 3.5 cm per Angstrom. The atomic and molecular orbital parts are represented by pastel/matte colored pieces and are color-coded according to their use. The pink and purple pear-shaped lobes and concave pi links represent the ve and -ve signs of the wave functions Psi of the lobes of atomic and molecular orbitals. The gray pieces represent hybridized sigma bonds. The light brown spheres are used to indicate lone electron pair orbitals. Where the electrons are involved in hydrogen bonding a light brown pear-shaped lobe is used
Catalog No. 50-290-1394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.