Learn More
United Scientific™ Moment of Inertia Kit
SDPSCI_ED
Catalog No. S000889 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Moment of Intertia Set
Catalog No. Product Type
S000889 Moment of Intertia Set
1 options
Prop 65
Catalog No. S000889 Supplier United Scientific™ Supplier No. MOMINRT
Add to Cart
Add to Cart
Prop 65
Demonstrate moment of inertia using this set of wands and rings.
The distribution of mass affects the moment of inertia of an object. The pair of wands both have the same mass and length. However, when the wand with a movable mass has the mass positioned near its end, the wand is much harder to move around than its counterpart which has its mass distributed evenly. The pair of rings also have similar mass and diameter. For the ring with movable masses, when the masses are positioned towards the center of the ring, it will roll very differently than when they are positioned near the outside edge. In both instances, with size and mass being the same, the only difference is the location of the center of mass. When an object is spinning, the farther away that the center of mass is from the point of rotation, the more effort it requires to move the object. Each of the wands has a mass of approximately 510 g and each of the rings has a mass of approximately 350 g.
  • Hands-on way to feel the moment of inertia
  • Sliding masses are easy to adjust
  • Includes activity guide
Specifications
Product Type Moment of Intertia Set