Learn More
New England Biolabs, Inc. Monarch® Protein Separation Solution - 36 ml
SDP
Supplier:  New England Biolabs, Inc. T3062L
Encompass
The Monarch Protein Separation Solution, supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue, is a salt-based solution that is optimized to ensure proteins in tissue and bacteria samples are easily removed during the phase separation of the Monarch HMW DNA Extraction workflow. It is supplied in two bottles of 18 ml.
  • Supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue
  • Facilitates the separation of proteins during HMW DNA extraction from tissues
  • Salt-based solution for protein precipitation
  • Supplied in 2 bottles of 18 ml
Catalog No. 50-206-8313