Learn More
New England Biolabs, Inc. Monarch RNA Purification Columns – 100 columns
SDP
Supplier:  New England Biolabs, Inc. T2007L
Encompass
Monarch RNA Purification Columns, supplied with the Monarch Total RNA Miniprep Kit, can purify up to 100 ug of total RNA from a variety of sample inputs. Columns are supplied with Monarch Collection Tubes II, and are compatible with vacuum manifolds.
Catalog No. 50-152-7885
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.