Learn More
Selleck Chemical LLC Monastrol 10mg 329689-23-8
SDP
Supplier:  Selleck Chemical LLC S843910MG
Encompass_Preferred
Monastrol is a cell-permeable small molecule inhibitor of kinesin-5(KIF11) which is essential for maintaining separation of the half-spindles. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.