Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey Complement C3a ELISA Kit (Colorimetric)
SDP

Catalog No. nbp270038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270038 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270038 Supplier Novus Biologicals™ Supplier No. NBP270038
Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Monkey Complement C3a ELISA Kit (Colorimetric) measures monkey Complement C3a in serum, plasma and other biological fluids.

Specifications

Assay Range 0.16 - 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Cross Reactivity This kit recognizes Monkey C3a in samples. No significant cross-reactivity or interference between Monkey C3a and analogues was observed.
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity Monkey Complement C3a ELISA Kit (Colorimetric) recognizes Monkey C3a in samples. No significant cross-reactivity or interference between Monkey C3a and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 718
Gene Symbol C3
Host Species Monkey
Quantity 1 Kit
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Neuroscience, Signal Transduction, Stem Cells
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Complement C3a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.