Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Monkey Diet, Standard Skf Diet, Powder

Diet for monkey research. Standard SKF.

$1049.00 - $3997.00

Specifications

Product Type Animal Diet
Autoclavable Not Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP290295110
View Documents
MP Biomedicals
0290295110
10 kg
Each for $1,049.00
Only null left
Add to Cart
 
MP290295120
View Documents
MP Biomedicals
0290295120
20 kg
Each for $1,831.00
Only null left
Add to Cart
 
MP290295150
View Documents
MP Biomedicals
0290295150
50 kg
Each for $3,997.00
Only null left
Add to Cart
 
Description

Description

Standard monkey diet containing casein, sucrose, fiber, salt mixture and vitamin fortified.

Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Not Autoclavable
Powder
Room Temperature
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.