Learn More
Vector Laboratories MONKEY F9/FACTOR IX ELISA KIT
SDP
Supplier:  Vector Laboratories LSF385351
Encompass
LS-F38535 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Monkey F9 / Factor IX in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of F9 / Factor IX as low as 0.19 nanograms per millilter.
Catalog No. NC2540957
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.