Learn More
Kingfisher Biotech Inc Squirrel Monkey IFN beta Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1829SM025
Encompass_Preferred
The Squirrel Monkey IFN beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Squirrel Monkey IFN beta applications are for cell culture, ELISA standard, and Western Blot Control. Squirrel Monkey IFN beta yeast-derived recombinant protein can be purchased in multiple sizes. Squirrel Monkey IFN beta Specifications: (Molecular Weight: 20.0 kDa) (Amino Acid Sequence: MSYNLLGFLQ RRSNFQSQKL LRQLNGRLEY CLKDRMDFEI PKEITQPQRL QKEDVALIVY EMLQNIFTIF NRDFSRTGWN KTIVENFLAN VYQHIEHLKT ILKEKWEKKE FTREKSMSSL HLKRYYGRIL HYLMAKEYSH CAWTIVSAEI LRNFYFIDRL TGYL (164)) (Gene ID: 101035681). For research use only. Made in the USA
Catalog No. 50-253-2722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.