Learn More
Rockland Immunochemicals Anti-MONKEY IgG (gamma chain) (GOAT) Antibody Peroxidase Conjugated - 617-103-012
SDP
Supplier:  Rockland Immunochemicals 617103012
Encompass_Preferred
Anti-Monkey IgG (gamma chain) peroxidase conjugated antibody generated in goat detects specifically Monkey IgG (gamma chain). This secondary peroxidase conjugated antibody anti-Monkey is ideal for investigators who routinely perform titration assays, western-blot, immunoprecipitation and more generally immunoassays.
Catalog No. NC0836125
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.