Learn More
MyBioSource Monkey SELE ELISA Kit
SDP
Supplier:  MyBioSource MBS250650196 TESTS
Encompass_Preferred
The MyBioSource Monkey SELE ELISA Kit quantifies SELE in various biological fluids using a 96-well plate with pre-coated antibodies, biotinylated antibodies, and HRP. It measures SELE levels from 15.6 to 1000 pg/mL.
Catalog No. NC2736318
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.