Learn More
American Research Products Inc Monkey thyroXine T4 ELISA ki
Supplier:  American Research Products Inc CSBEQ027512MK
Encompass
Monkey thyroXine (T4) ELISA kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27271