Learn More
Vector Laboratories MONKEY TTR /TRANSTHYRETIN KIT
SDP
Supplier:  Vector Laboratories LSF492581
Encompass
Monkey TTR / Transthyretin ELISA Kit (Sandwich ELISA).1 Plate.96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Monkey TTR / Transthyretin in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of TTR / Transthyretin as low as 0.19 micrograms per millilter.
Catalog No. NC1744159
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.