Learn More
Sigma Organic Chemistry mono-N-Dodecyl phosphat | 250mg | 2627-35-2 | MFCD00013946
Supplier: Sigma Organic Chemistry CDS001542250MG

mono-N-Dodecyl phosphat, 250mg
About this Item:
CAS #: 2627-35-2
MDL #: MFCD00013946
Molecular Weight: 266.31
UNSPSC Code: 12352200
Chemical Formula: C12H27O4P
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.