Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Mono Q PC, Columns Prepacked with MonoBeads™

Catalog No. p-3676480 Shop All Cytiva Products
Change view
Click to view available options
Height (Metric):
100mm
Material:
PEEK
Max. Pressure:
40 bar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Material Max. Pressure
45-000-309 100mm PEEK 40 bar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-309 Supplier Cytiva Supplier No. 17517901

Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. Cytiva Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.

  • Maximum resolution achieved from efficiency of small spherical particles, optimally packed
  • Particles are coupled to Q or S ion exchangers with excellent sensitivities
  • Compatible with high-performance LC systems
  • Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid

  • Tricorn columns are optimized for the polishing step when high purity is required
  • PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
  • Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
  • Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Product Type Mono Q ion exchange column
Bed Volume 1.7mL
Diameter (English) Inner 0.18 in.
Diameter (Metric) Inner 4.6mm
Quantity Each
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate 0.05 to 3mL/min.
Material PEEK
Ligand Type O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3
Max. Pressure 40 bar
pH Range 2 to 12
Particle Size 10μm
Support Type Monodisperse porous Polystyrene/Divinyl Benzene bead
Content And Storage 4° to 30°C
Column Type Mono Q Ion Exchange Column
For Use With (Application) Small-scale polishing in purification of Proteins, Peptides and other biomolecules when high purity is required
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.