Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Mono S HR 16/10 Prepacked Column

Catalog No. 45002553 Shop All Cytiva Products
Change view
Click to view available options
Column prepacked with Mono S HR 16/10 media
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
45-002-553 Column prepacked with Mono S HR 16/10 media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-553 Supplier Cytiva Supplier No. 17050701

missing translation for 'validMainframeMsg'

Specifications

Bed Volume 20 mL
Flow Rate less than 10 mL/min.
Content And Storage 4° to 30°C, in 20% Ethanol
Column Type Mono S Ion Exchange Column
For Use With (Application) Ion Exchange Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.