Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobasic Potassium Phosphate, Crystal, NF, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604653 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-653 2.5 kg Poly Bottle
18-604-652 6 x 500 g Poly Bottle
18-604-654 4 x 2.5 kg Poly Bottle
18-604-655 12 kg Poly Pail
18-604-656 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-653 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO2002.5KG
Add to Cart
Add to Cart

PO200, 7778-77-0, KH2PO4

Spectrum™ Chemical Monobasic Potassium Phosphate, Crystal, NF is used as a buffering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade NF
Loss on Drying 0.01
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.