Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal ANTI-FLAG M2 antibody produced in mouse, 5 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F18045MG
Encompass
For highly sensitive and specific detection of FLAG fusion proteins by immunoblotting, immunoprecipitation (IP) , immunohistochemisty, immunofluorescence and immunocyotchemistry. Optimized for single banded detection of FLAG fusion proteins in mammalian, plant, and bacterial expression systems.
Catalog No. NC1895991
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.