Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti Vinculin a
Supplier:  Sigma Aldrich Fine Chemicals Biosciences V4505.2ML
Encompass
The vinculin (VCL) gene containing 22 exons is mapped to human chromosome 10q22.1-q23. The gene codes for a 117kDa cytoskeletal protein present in adherens type cell junctions. VCL is a ubiquitously expressed cell-adhesion protein. The protein is characterized with a globular head and a tail domain which are interconnected by a short proline-rich sequence an intramolecular interaction between the head and tail conceal many ligand-binding sites in the protein.
Catalog No. 50-290-5428
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.