Learn More
Cell Signaling Technology Phospho-S6 Ribosomal Protein (Ser235/236) (D57.2.2E) XP(R) Rabbit mAb (Alexa Fluor(R) 647 Conjugate) 100 ul
SDP
Supplier:  Cell Signaling Technology 4851S
Encompass
4851S , One way that growth factors and mitogens effectively promote sustained cell growth and proliferation is by upregulating mRNA translation (1,2). Growth factors and mitogens induce the activation of p70 S6 kinase and the subsequent phosphorylation of the S6 ribosomal protein. Phosphorylation of S6 ribosomal protein correlates with an increase in translation of mRNA transcripts that contain an oligopyrimidine tract in their 5' untranslated regions (2). These particular mRNA transcripts (5'TOP) encode proteins involved in cell cycle progression, as well as ribosomal proteins and elongation factors necessary for translation (2,3). Important S6 ribosomal protein phosphorylation sites include several residues (Ser235, Ser236, Ser240, and Ser244) located within a small, carboxy-terminal region of the S6 protein (4,5).
Catalog No. 50-191-391