Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethanolamine, NF, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604682 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-682 200 L Poly Drum
18-604-680 4 L Amber Glass Bottle
18-604-681 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-682 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1322200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1322, 141-43-5, C2H7NO

Spectrum™ Chemical Monoethanolamine, NF - All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

CAS Max % 1
Boiling Point 170.8°C / 339.4°F
Ignition Residue 0.001
Packaging Poly Drum
Quantity 200 L
Percent Purity 98 to 100.5%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Monoethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.