CAS RN 141-43-5
2-Aminoethanol 99.0+%, TCI America™
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Ethanolamine, 99%
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 SMILES: NCCO
Ethanolamine, ACS reagent
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 SMILES: NCCO
Ethanolamine, 99%, AcroSeal™
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 SMILES: NCCO
VeriSpec™ Monoethanolamine -H3NC2H4OH+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Ethanolamine, 99%, H2O 0.5% max, Thermo Scientific Chemicals
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethanol SMILES: NCCO
Monoethanolamine, NF, 98-100.5%, Spectrum™ Chemical
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Ethanolamine, ACS, 99+%
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 SMILES: NCCO
Monoethanolamine, NF, 98-100.5%, Spectrum™ Chemical
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Ethanolamine, 98+%
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N Synonym: ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine PubChem CID: 700 ChEBI: CHEBI:16000 SMILES: NCCO
2-Aminoethanol, ACS, 99%, Spectrum™ Chemical
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N IUPAC Name: 2-aminoethan-1-ol SMILES: NCCO
Ethanolamine, SPEX CertiPrep™
CAS: 141-43-5 Molecular Formula: C2H7NO Molecular Weight (g/mol): 61.08 MDL Number: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethanol SMILES: NCCO
Sigma Aldrich Ethanolamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2