Learn More
U.S. Pharmacopeia Monoglycerides, 68990-53-4, 125mg
Supplier: U.S. Pharmacopeia 1446000
Melting Point 158 °F (70 °C), Flash Point 496.40 °F (258.00 °C) Cleveland Open Cup
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.