Learn More
Covidien Monoject™ Softpack Hypodermic Needles with Polypropylene Hub
Description
- Ultra-sharp tri-beveled surgical grade stainless steel coated with a special grade of silicone produces a consistently sharp needle that reduces both the force needed to penetrate and any “drag” on the injection site, producing more comfortable injections for the patient
- Needles are autoclavable, allowing convenient use in hospital-prepared surgical packs
- Cap-to-sheath seal provides visible and audible tamper evidence and guarantees sterility
- Rigid polypropylene packaging protects against accidental product contamination
- Easy-grip design allows easy opening of packaging
Bevels available include: Regular (A) for intramuscular and subcutaneous injections, Intradermal or Intermediate (IN) for intradermal injections, and Short (B) for intravenous or intrarterial applications and nerve blocks
Specifications
Specifications
| Material | Polypropylene (Hub), Stainless Steel |
| Needle Gauge | 21 G |
| Latex Free | Yes |
| Length (English) Needle | 1.5 in. |
| Product Type | Monoject™ Replacement Hypodermic Needle |
| Needle Point Style | Regular Bevel |
| For Use With (Equipment) | Monoject Syringe |
| Sterility | Sterile |