Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl Fumarate 98.0+%, TCI America™
SDP

Catalog No. M241325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M241325G 25 g
M24135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M241325G Supplier TCI America Supplier No. M241325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2756-87-8
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.099
MDL Number MFCD00063174
InChI Key NKHAVTQWNUWKEO-NSCUHMNNSA-N
Synonym monomethyl fumarate, 4-methoxy-4-oxobut-2-enoic acid, methyl hydrogen fumarate, mono-methyl fumarate, fumaric acid monomethyl ester, unii-45iub1px8r, monomethylfumarate, 45iub1px8r, e-4-methoxy-4-oxobut-2-enoic acid, 2e-4-methoxy-4-oxobut-2-enoic acid
PubChem CID 5369209
IUPAC Name (E)-4-methoxy-4-oxobut-2-enoic acid
SMILES COC(=O)C=CC(=O)O

Specifications

Melting Point 147°C
Color White
Quantity 25 g
Formula Weight 130.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Fumarate
EINECSNumber (2)-1142
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.