Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl Itaconate 98.0+%, TCI America™
SDP

Catalog No. I0269100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0269100G 100 g
I026925G 25 g
I0269500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0269100G Supplier TCI America Supplier No. I0269100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7338-27-4
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00021713
InChI Key OIYTYGOUZOARSH-UHFFFAOYSA-N
Synonym Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate
PubChem CID 81791
IUPAC Name 4-methoxy-2-methylidene-4-oxobutanoic acid
SMILES COC(=O)CC(=C)C(=O)O

Specifications

Melting Point 72°C
Color White
Boiling Point 149°C
Quantity 100 g
Formula Weight 144.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Itaconate
EINECSNumber (2)-2615
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.