Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate 98.0+%, TCI America™
SDP

Catalog No. A033725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033725G 25 g
A0337500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A033725G Supplier TCI America Supplier No. A033725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 842-15-9
Molecular Formula C10H9KNO6S2
Molecular Weight (g/mol) 342.401
MDL Number MFCD00064177
InChI Key QNHMTLCVDUMUFH-UHFFFAOYSA-N
Synonym Amino G Acid Monopotassium Salt, 7-Amino-1,3-naphthalenedisulfonic Acid Monopotassium Salt, 2-Naphthylamine-6,8-disulfonic Acid Monopotassium Salt, Potassium Hydrogen 7-Amino-1,3-naphthalenedisulfonate
PubChem CID 88146376
IUPAC Name 7-aminonaphthalene-1,3-disulfonic acid;potassium
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N.[K]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 341.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.