Learn More
Genscript Corporation MonoRab Anti-DYKDDDDK Magnetic Beads, 70 μm average bead size, >1.5 mg binding capacity per 1 ml, reusable ≥5 times, stored at -20 °C, for protein purification applications
Supplier:  Genscript Corporation L008355
Encompass
MonoRab Anti-DYKDDDDK Magnetic Beads, 70 μm average bead size, >1.5 mg binding capacity per 1 ml, reusable ≥5 times, stored at -20 °C, for protein purification applications
Catalog No. NC2481266
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.