Learn More
Genscript Corporation D-dimer 15C18 mAb Mouse
Supplier:  Genscript Corporation V014031
Encompass_Preferred
D-dimer known as a fibrin degradation product is presented in blood after a blood clot is degraded by fibrinolysis Its concentration in blood increases when deep venous thrombosis (DVT) pulmonary embolism (PE) or disseminated intravascular coagulation (DIC) happens It serves as a useful maker for the diagnosis of these diseases
Catalog No. 50-000-56798
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.