Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Humanized VHH Antibody [iFluor 647], mAb
Supplier:  Genscript Corporation A02170200
Encompass_Preferred
The product is specific for Humanized VHH antibodies and Camelid (Llama, Alpaca, and Camel) heavy chain antibodies (IgG2&3), but does not recognize the camelid conventional antibody (IgG1).
Catalog No. NC2137110
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.