Learn More
Genscript Corporation Antibody Beads Other Products Immunodetection 1 ML 1/EA
Supplier:  Genscript Corporation L01008
Encompass
MonoRab Rabbit Anti-VHH Microbeads are coated with anti-VHH antibodies and can be used to separate VHH+ cells from fresh or frozen camelid peripheral blood mononuclear cells (PBMCs) or single cell suspensions based on the surface expression of VHH.
Catalog No. 50-252-9969
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.