Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monosodium L-Glutamate, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604684 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-684 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-684 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G1038500GM

missing translation for 'validMainframeMsg'

G1038, 6106-04-3, C5H8NNaO4·H2O

Spectrum™ Chemical Monosodium L-Glutamate, FCC is used in the food industry as a flavor with an umami taste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Molecular Weight (g/mol) 187.13
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
IUPAC Name sodium (4S)-4-amino-4-carboxybutanoate hydrate
SMILES O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O

Specifications

CAS Max % 1
Lead (Pb) 5 mg/kg
Loss on Drying 0.005
Packaging Poly Bottle
Quantity 500 g
Optical Rotation +24.5° to+25.5°
Percent Purity 98.5 to 101.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.