Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Montage™ Plasmid Miniprep96 Kits

Catalog No. LSKP09604 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Reactions:
4 x 96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
LSKP09604 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKP09604 Supplier MilliporeSigma™ Supplier No. LSKP09604
Add to Cart
Add to Cart

Includes

All necessary reagents and disposables

Requires

MultiScreen™ Vacuum Manifold with underdrain support grid (MAVM 096 0R)

Generate high-quality plasmid DNA in 50% less time than traditional methods

  • Typical yield from 1mL of bacterial culture is 4 to 6μg
  • 96-well kit uses unique separation technology Simple, 25-minute protocol yields clean and reproducible DNA for sensitive downstream applications Collection of purified sample from the top of the plate and lack of centrifugation steps allow compatibility with automated systems

TRUSTED_SUSTAINABILITY

Specifications

Format Microplate
For Use With (Application) Plasmid Miniprep, BAC Prep
Includes All necessary reagents and disposables
No. of Reactions 4 x 96 Preps
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.