Learn More
Apexbio Technology LLC Montelukast,100mg CAS# 158966-92-8
SDP
Supplier:  Apexbio Technology LLC C6416100
Encompass_Preferred
Montelukast is a potent, selective and orally-active antagonist of the cysteinyl leukotriene 1 (CysLT1) receptor, with an IC50 value of 4.9 nM in HEK293 cell membranes expressing human CysLT1 receptors. The CysLT1 receptor is a receptor for cysteinyl leukotrienes (e.g. LTC4, LTD4, and LTE4) which mediate allergic and hypersensitivity reactions in a variety of inflammatory disorders, in particular asthma. Montelukast has been introduced clinically for the treatment of asthma, allergic rhinitis, and exercise-induced bronchoconstriction.  Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0574
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.